Distilling receivers, Bernauer
Distilling receivers, Bernauer
19/26 NS
N° de catalogue LENZ05288102
Fournisseur:   Lenz Laborglas
Distilling receivers, Bernauer
N° de catalogue LENZ05288102
Fournisseur:  Lenz Laborglas
Distilling receivers, Bernauer
Distilling receivers, Bernauer
19/26 NS
Prix
92,90 €
Connexion Pour la disponibilité
1 * 1 ST
Product Details & Documents
DURAN®, borosilicate glass, clear.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
Distilling receivers acc. to Bernauer (modified), with three cones NS 14/23, vacuum connection with glass olive. With flasks 100 ml capacity or without flasks available.
Documents
Spécifications
  • Rodage femelle:
    19/26 NS