Erlenmeyer flasks, narrow neck, with baffles
Erlenmeyer flasks, narrow neck, with baffles
1000 ml
N° de catalogue 214-0187
Erlenmeyer flasks, narrow neck, with baffles
N° de catalogue 214-0187
Erlenmeyer flasks, narrow neck, with baffles
1000 ml
Prix
118,00 €
Connexion Pour la disponibilité
1 * 1 ST
Product Details & Documents
Borosilicate glass 3.3.
  • Excellent chemical resistance
  • High temperature resistance
  • Minimal thermal expansion, giving relatively high resistance to temperature changes
Ideal for use in the preparation of cultures.
Spécifications
  • Capacité:
    1000 ml
  • Seq:
    10