Erlenmeyer flasks, narrow neck, with baffles
500 ml
214-0186
Borosilicate glass 3.3.
- Excellent chemical resistance
- High temperature resistance
- Minimal thermal expansion, giving relatively high resistance to temperature changes
Ideal for use in the preparation of cultures.
- Capacité:500 ml
- Seq:09