Erlenmeyer flasks, narrow neck, with baffles
1000 ml
N° de catalogue 214-0183
Fournisseur: NEUBERT VOLUME GLASSWAERE
Erlenmeyer flasks, narrow neck, with baffles
N° de catalogue 214-0183
Fournisseur: NEUBERT VOLUME GLASSWAERE
Aller à :
Product Details & Documents
Borosilicate glass 3.3.
- Excellent chemical resistance
- High temperature resistance
- Minimal thermal expansion, giving relatively high resistance to temperature changes
Ideal for use in the preparation of cultures.
Spécifications
- Capacité:1000 ml
- Seq:04