Fioles à vide forme erlenmeyer
500 ml
N° de catalogue 511-0003
Fournisseur: NEUBERT VOLUME GLASSWAERE
Aller à :
Product Details & Documents
DURAN®, borosilicate glass, plastic coated.
- For vacuum use
- Does not comply with recent German safety regulations
- Olive hose connection 11 mm Ø
The coating provides scratch, leak and splinter protection.
Spécifications
- Capacité:500 ml



