ContactUsLinkComponent
TUBE COLORIMETRIQUE SELON NESSLER, NEUBERT, FORME BASSE, DIVISION (ML): 100, CAPACITE (ML): 100, X L (MM): 26 X 320 1 * 1 Pce
  NEUB0387-32-222
TUBE COLORIMETRIQUE SELON NESSLER, NEUBERT, FORME BASSE, DIVISION (ML): 100, CAPACITE (ML): 100, X L (MM): 26 X 320 1 * 1 Pce
  NEUB0387-32-222
Frequently Bought Together