Oxygen/moisture traps
Oxygen/moisture trap, stainless steel, with fitting size ¼", 15 ppb
N° de catalogue HEWLOT3-4-SS
Fournisseur: Agilent
Aller à :
Product Details & Documents
Oxygen/moisture traps have two functionalities in the same trap. These traps are disposable. In addition to inert gases (nitrogen, helium, argon and krypton), the Agilent OT3 trap treats streams of hydrogen, alkanes, alkenes, aliphatic hydrocarbon gases, low boiling aromatics, carbon dioxide, carbon monoxide, and argon-methane.
- Leak-free, one piece design
- Bed material treated with ultra high purity helium
- Filter design prevents channeling, promotes efficient scrubbing
The OT3 trap contains a highly active, metal containing, scrubbing material in an inert, aluminum body. This material removes 500 ml oxygen from a gas stream to less than 15 ppb. Additionally, up to 2 grams of moisture can be removed by this trap without affecting the oxygen capacity.
Spécifications
- Désignation:Oxygen/moisture trap, stainless steel, with fitting size ¼", 15 ppb
- Seq:04