Saisie de commande
Belgium
Lien pour commandesContactUsLinkComponent
Pepstatin A

Pepstatin A

Flacon verre pour solides, 100 mg
Catalog # BMOL17640-2
Pepstatin A
Catalog # BMOL17640-2
CAS Number:  
Pepstatin A

Flacon verre pour solides, 100 mg

CAS Number:  
Price
Inquire For Price
Log in for availability
1 * 100 mg
Product Details & Documents
Specifications

Specifications

  • Type de conditionnement:
    Flacon verre pour solides
  • Cdt:
    100 mg
  • Cat. No.:
    BMOL17640-2
  • N ° CAS.:
    26305-03-3
  • Conditions de stockage:
    Refrigerator
  • Molecular formula:
    C₃₄H₆₃N₅O₉
  • MDL No.:
    MFCD00060740
  • Nom de la protéine/du peptide:
    Pepstatin A

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.