Glass Adapters
Product Details & Documents
ROBU® Glass Adapter for Crucibles is designed to securely connect filter crucibles to other laboratory apparatus. Made from durable glass, it ensures a leak-free fit, offering reliable and efficient performance during filtration and chemical processes.
- Leak-free connection
- Durable glass construction
- Resistant to heat and chemicals
- Ensures secure attachment for crucibles
- Compatible with various filtration setups
- Easy to clean and reusable
- Reliable performance in laboratory filtration processes
The ROBU Glass Adapter for Crucibles is designed to provide a secure and leak-free connection between filter crucibles and other laboratory apparatus. Constructed from durable, heat-resistant glass, this adapter is highly resistant to chemicals and ensures consistent, reliable performance during filtration processes. It is compatible with a wide range of filtration setups, offering versatility and ease of use in various laboratory applications. The adapter is easy to clean and reusable, making it a cost-effective solution for maintaining high standards in laboratory filtration work.
Certifications: In compliance with ISO 4796-80 and ASTM E128-99 standards.
Documents
TDS VitraPOR Borosilicate Glass 3.3.pdf
Fiche technique
Spécifications
- Ø ext.:10 mm
- Désignation:Glass adapter; inner tube Ø: 40 mm; outer tube Ø: 10 mm
- Matière:Verre borosilicaté 3.3
- Seq:3
Les recommandations seront personnalisées en fonction de vos préférences d’achat uniquement si vous avez donné votre consentement en activant l’option " Améliorer mon expérience d’achat " sur la page "Mon compte".
Dans le cas contraire, vous recevrez des recommandations génériques.



