Evaporating flasks
50 ml, 29/42 NS, uncoated
N° de catalogue BUCH008736
Fournisseur: BUCHI
Aller à :
Product Details & Documents
Borosilicate glass.
- Very good chemical resistance
- High temperature resistance
- Minimal thermal expansion, giving relatively high resistance to temperature changes
For Rotavapor® rotary evaporators. Centre neck. Pear-shaped with ground joint socket or EL-flange.
Spécifications
- Rodage femelle:29/42 NS
- Type:uncoated
- Capacité:50 ml
- Seq:03