Spider evaporators
Spider evaporators
Spider with cylindric flasks, SJ29/32, glass, incl. 6 cylindric flasks, SJ14.5/23, 20 ml, For simultaneous distillation from 6×20 ml cylindrical flasks with 29/32 NS Joint
N° de catalogue 531-1321
Fournisseur:   BUCHI
Spider evaporators
N° de catalogue 531-1321
Fournisseur:  BUCHI
Spider evaporators
Spider evaporators
Spider with cylindric flasks, SJ29/32, glass, incl. 6 cylindric flasks, SJ14.5/23, 20 ml, For simultaneous distillation from 6×20 ml cylindrical flasks with 29/32 NS Joint
Prix
1 760,00 €
Connexion Pour la disponibilité
1 * 1 ST
Product Details & Documents
Spider evaporators are ideal when users need to concentrate several samples under the same conditions.
  • Models for distillation with evaporating or cylindrical flasks available
  • Parallel distillation saves time
Spécifications
  • Désignation:
    Spider with cylindric flasks, SJ29/32, glass, incl. 6 cylindric flasks, SJ14.5/23, 20 ml
  • Type:
    For simultaneous distillation from 6×20 ml cylindrical flasks with 29/32 NS Joint