Plaques
Plaques
Métal, 285 mm
Catalog # 467-1931
Supplier:   DWK Life Sciences
Plaques
Catalog # 467-1931
Supplier:  DWK Life Sciences
Plaques
Plaques
Métal, 285 mm
Price
55,70 €
Log in for availability
1 * 1 ST
Product Details & Documents
Stainless steel 1.4301, Type 304, rust-free.
Specifications
  • Ø:
    285 mm
  • Type:
    Métal
  • Seq:
    07