Béchers, forme basse
500 ml, , 110 mm, 80 mm
N° de catalogue BOCH8532
Fournisseur: BOCHEM
Aller à :
Product Details & Documents
Stainless steel, 18/10.
- Good chemical resistance
- Excellent resistance to heat
- Smooth surfaces for fast, easy cleaning
Ideal for corrosive materials. Robust and durable.
Spécifications
- Ø ext.:80 mm
- Capacité:500 ml
- Hauteur:110 mm