Attaches, micro
Clamps, micro, finger with cork, angled jaws, Acier inoxydable 18/10, 15 mm, 150 mm
N° de catalogue BOCH5405
Fournisseur: BOCHEM
Aller à :
Product Details & Documents
Micro clamps made of stainless steel.
- Fingers are covered with either a layer of cork or with silicone coating to protect clamped items
- With round or angled jaws
Spécifications
- Largeur de serrage:15 mm
- Désignation:Clamps, micro, finger with cork, angled jaws
- Longueur:150 mm
- Matière:Acier inoxydable 18/10