Pressure Bulbs
Pressure Bulbs
Ball set with net, black, size 5, natural rubber/PVC, 102
N° de catalogue 612-9953
Fournisseur:   DEUTSCH NEUMANN
Pressure Bulbs
N° de catalogue 612-9953
Fournisseur:  DEUTSCH NEUMANN
Pressure Bulbs
Pressure Bulbs
Ball set with net, black, size 5, natural rubber/PVC, 102
Prix
107,00 €
Connexion Pour la disponibilité
1 * 1 ST
Product Details & Documents
Natural rubber, chloroprene or PVC.
Used for filling automatic burettes, Pellet pattern (ball set with net) or atomisers (bulb).
Spécifications
  • Capacité:
    102
  • Désignation:
    Ball set with net, black, size 5, natural rubber/PVC
  • Seq:
    4