Double bossheads
Double bosshead, with safety screw type 'heart' M8, 16,5 mm
451-9111
: BOCHEM
Double bossheads made of stainless steel 18/10.
- For rods up to 16,5 mm
- Angle: 90°
- Available with heart screw M6/M8, socket screw M6/M8 or safety screw type 'heart' M8
: DIN 12895
- Désignation:Double bosshead, with safety screw type 'heart' M8
- Pour tige Ø:16,5 mm
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



