Distributeurs, close-it
Distributeur d'étiquettes de contrôle 95×95 mm
Catalog # 312-1156
Supplier: BURKLE
Jump to:
Product Details & Documents
Dispensers for control seals labels.
- Powerful strength of the adhesive
- Two sizes are available
- Reliably prevents products from leaking out after sampling
Particularly suited for the food and pharma industries.
Certifications: FDA guidelines.
Ordering information: Two different sizes available.
Documents
BURK5303-9000
Images 360
Specifications
- Désignation:Distributeur d'étiquettes de contrôle 95×95 mm
- Seq:901