Fused quartz microscope slides
Microscope slides, Verre de quartz, 26×51 mm
Fournisseur: Thermo Fisher Scientific
Aller à :
Product Details & Documents
These fused quartz slides are made of fusing naturally occurring crystalline silica.
- High chemical purity, high corrosion resistance, high melting point
- Low coefficient of thermal expansion with excellent electrical insulation
- Optical transmission from ultra-violet to infrared
Documents
Spécifications
- Désignation:Microscope slides
- L×l:26×51 mm
- Matière:Verre de quartz
- Seq:05