Autoclavable baskets
470×300×200 mm
Fournisseur: DELTALAB
Aller à :
Product Details & Documents
Made of stainless steel wire.
- Stackable
- Good chemical resistance
- Excellent temperature resistance
Baskets for autoclaves and carrying.
Documents
Certificats
Rechercher par numéro de lotSpécifications
- Seq:01
- l×p×h:470×300×200 mm