Infrared optical windows
Crystal optic disc, polished both sides, Silver chloride, 13 mm, 2 mm
These optical crystal windows are ideal for infrared spectroscopy.
- Disc or rectangular
- Polished or unpolished
A polishing kit is available for those who wish to polish their windows.
- Matière fenêtre:Silver chloride
- Désignation:Crystal optic disc, polished both sides
- Dimensions:13 mm
- Seq:80
- Épaisseur:2 mm



