Saisie de commande
Belgium
Lien pour commandesContactUsLinkComponent
 

Dichlorométhane ≥99.5% stabilisé ACS

Flacon Verre, 500 mL
Catalog # 39116.AP
Health
Irritant
Dichlorométhane ≥99.5% stabilisé ACS
Catalog # 39116.AP
CAS Number:  75-09-2
Dichlorométhane ≥99.5% stabilisé ACS
Dichlorométhane ≥99.5% stabilisé ACS

Flacon Verre, 500 mL

CAS Number:  75-09-2
Price
37,90 €
Log in for availability
1 * 500 mL
Product Details & Documents

Purity: 99.5+%
CAS No.: 75-09-2
Molecular Formula: CH2Cl2
Formula Weight: 84.93
Physical Form: Liquid
Density: 1.325
Refractive Index: 1.4244
Appearance: Colourless
Odour: Sweet
Melting Point: -95°
Boiling Point: 39-40°
Beilstein Reference: 1730800
Merck Reference: 14,6063
UN No.: 1593
MDL No.: MFCD00000881

Synonyms:  Methylene chloride, DCM
  • CAS Number:75-09-2
  • Formula:CH₂Cl₂
  • MDL Number:MFCD00000881
  • Molecular Weight:84.93 g/mol
  • Merck Index:14,6063
  • Boiling Pt:39…40 °C
  • Melting Pt:–95 °C
  • Density:1,325
  • Storage Temperature:Température ambiante
  • EINECS:200-838-9
  • ADR:6.1,III
  •  :Stabilisé avec de l'Amylène
  • UN:1593
Specifications

Specifications

  • Type de conditionnement:
    Flacon Verre
  • Cdt:
    500 mL

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.