Infrared optical windows
Crystal optic disc, polished both sides, Calcium fluoride, 13 mm, 1 mm
Fournisseur: Thermo Fisher Scientific
Aller à :
Product Details & Documents
These optical crystal windows are ideal for infrared spectroscopy.
- Disc or rectangular
- Polished or unpolished
A polishing kit is available for those who wish to polish their windows.
Spécifications
- Matière fenêtre:Calcium fluoride
- Désignation:Crystal optic disc, polished both sides
- Dimensions:13 mm
- Seq:48
- Épaisseur:1 mm
Accessories
Informations sur le produit
- N° de catalogueDésignationDisponibilitéPrix



