Saisie de commande
Belgium
Lien pour commandesContactUsLinkComponent
Sodium hydroxyde 50% (p/p) en solution aqueuse

Sodium hydroxyde 50% (p/p) en solution aqueuse

Flacon plastique, 5 kg
Catalog # 33382.A7
Corrosive
Sodium hydroxyde 50% (p/p) en solution aqueuse
Catalog # 33382.A7
CAS Number:  1310-73-2
Sodium hydroxyde 50% (p/p) en solution aqueuse
Sodium hydroxyde 50% (p/p) en solution aqueuse

Flacon plastique, 5 kg

CAS Number:  1310-73-2
Price
214,00 €
Log in for availability
1 * 5 kg
Product Details & Documents

CAS No.: 1310-73-2 Molecular Formula: NaOH Formula Weight: 40.00 Physical Form: Liquid Density: 1.53 Appearance: Colourless UN No.: 1824 MDL No.: MFCD00003548

Synonyms:  Caustic soda
  • CAS Number:1310-73-2
  • Formula:NaOH
  • MDL Number:MFCD00003548
  • Molecular Weight:40 g/mol
  • Boiling Pt:119…135 °C (1013 hPa)
  • Melting Pt:9…10 °C
  • Density:1,53
  • Storage Temperature:Température ambiante
  • EINECS:215-185-5
  • UN:1824
  • ADR:8,II
Specifications

Specifications

  • Type de conditionnement:
    Flacon plastique
  • Cdt:
    5 kg

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.