Acétone ≥99.5% ACS
Flacon Verre, 1 L
Fournisseur: Thermo Fisher Scientific
Acétone ≥99.5% ACS
N° de catalogue 30698.K2
Fournisseur: Thermo Fisher Scientific
Numéro CAS: 67-64-1
Aller à :
Product Details & Documents
Purity: 99.5+% CAS No.: 67-64-1 Molecular Formula: C3H6O Formula Weight: 58.08 Flash Point: -17°(1°F) Physical Form: Liquid Density: 0.791 Refractive Index: 1.3590 Appearance: Colourless Odour: Characteristic Melting Point: -94° Boiling Point: 56° Beilstein Reference: 635680 Merck Reference: 14,66 UN No.: 1090 MDL No.: MFCD00008765
Synonymes: 2-Propanone, DMK, Propanone, Dimethyl ketone
- Numéro CAS:67-64-1
- Formule:CH₃COCH₃
- Numéro MDL:MFCD00008765
- Poids moléculaire:58.08 g/mol
- Index Merck:14,66
- Point d'ébullition:56 °C
- Point de fusion:–94 °C
- Densité:0,791
- Point d'éclair:–17 °C (1 °F)
- Température de stockage:Température ambiante
- EINECS:200-662-2
- UN:1090
- ADR:3,II
- REACH:01-2119471330-49
Documents
Spécifications
Spécifications
- Type de conditionnement:Flacon Verre
- Cdt:1 L
- Cat. No.:30698.K2
- Certification environnementale:Chem21 Solvent Selection Guide recommended after discussion
- Environmentally Preferable:Safer Human and Environmental Health
Spécifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



