Saisie de commande
Belgium
Lien pour commandesContactUsLinkComponent
Evaporating dishes, high shape

Evaporating dishes, high shape

1000 ml, 75 mm, 140 mm
N° de catalogue BOCH8563
Fournisseur:   BOCHEM
Evaporating dishes, high shape
N° de catalogue BOCH8563
Fournisseur:  BOCHEM
Evaporating dishes, high shape
Evaporating dishes, high shape

1000 ml, 75 mm, 140 mm

Prix
87,60 €
Connexion Pour la disponibilité
1 * 1 ST
Product Details & Documents

Evaporating dish, stainless steel, with contact bottom for heating plates and magnetic stirrers.

  • Very good temperature resistance
  • Good chemical resistance
  • Easy to clean
Spécifications
  • Ø ext.:
    140 mm
  • Capacité:
    1000 ml
  • Hauteur:
    75 mm