Cheers, tray
Cheers, tray
288×428×17 mm
N° de catalogue VITL71998
Fournisseur:   VITLAB
Cheers, tray
N° de catalogue VITL71998
Fournisseur:  VITLAB
Cheers, tray
Cheers, tray
288×428×17 mm
Prix
58,80 €
Connexion Pour la disponibilité
1 * 1 ST
Product Details & Documents
Made of melamine, white.
  • Rounded corners
  • Smooth surfaces, easy to clean
  • Stable and self-supporting
Practical tray for instruments, tools, and sensitive utensils.
Spécifications
  • l×p×h:
    288×428×17 mm