Double-spatules, étroites
Rigide, 150 mm, 9 mm
Catalog # 231-0563
Supplier: RSG
Jump to:
Product Details & Documents
Double-ended, narrow spatulas made from stainless steel. Available flexible or non flexibe.
Specifications
- Désignation:Rigide
- Longueur:150 mm
- Largeur de la lame:9 mm