Restore™ Western Blot Stripping Buffer
Restore™ Western Blot Stripping Buffer, sufficient for stripping 25 blots (8×10 cm), 500 mL
N° de catalogue PIER21059
Fournisseur: Thermo Fisher Scientific
Restore™ Western Blot Stripping Buffer
N° de catalogue PIER21059
Fournisseur: Thermo Fisher Scientific
Numéro CAS:
Aller à :
Product Details & Documents
Although optimising assay conditions is the best way to achieve optimum results, re-performing the gel electrophoresis process to test each new primary antibody or antibody concentration is time-consuming and expensive. Restore™ Western blot stripping buffer offers an effective alternative.
- Saves time - no need to re-run gels
- Saves precious sample
- Provides efficient removal
- Gentle formulation ensures target protein is not damaged after stripping and re-probing
- Odour-free
Spécifications
Spécifications
- Cdt:500 mL
- Désignation:Restore™ Western Blot Stripping Buffer, sufficient for stripping 25 blots (8×10 cm)
Spécifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



