Reducing pieces with small flanges KF
Reducing piece, DN 25/10, Aluminium
Catalog # 185-2301
Supplier: VACUUBRAND
Jump to:
Product Details & Documents
Reducing pieces with small flanges KF made of aluminium or stainless steel.
Documents
MB_Reduzierstuecke_KF
Technical Drawing
Specifications
- Désignation:Reducing piece, DN 25/10
- Matière:Aluminium
- Seq:01