Small flanges with thread
Small flanges with thread
Small flanges KF, NW 16 with thread, G1/4", for ME 2, MZ 2, MV 2, DN 16
N° de catalogue 185-2277
Fournisseur:   VACUUBRAND
Small flanges with thread
N° de catalogue 185-2277
Fournisseur:  VACUUBRAND
Small flanges with thread
Small flanges with thread
Small flanges KF, NW 16 with thread, G1/4", for ME 2, MZ 2, MV 2, DN 16
Prix
41,00 €
Connexion Pour la disponibilité
1 * 1 ST
Product Details & Documents
These small flanges with thread made of stainless steel.
Documents
Spécifications
  • Désignation:
    Small flanges KF, NW 16 with thread, G1/4", for ME 2, MZ 2, MV 2, DN 16
  • Seq:
    01