Closures for headspace vials
PTFE coated silicone cap, pre-assembled
N° de catalogue PERCB4000022
Fournisseur:   PerkinElmer
Closures for headspace vials
N° de catalogue PERCB4000022
Fournisseur:  PerkinElmer
Closures for headspace vials
PTFE coated silicone cap, pre-assembled
Prix
1 300,00 €
Connexion Pour la disponibilité
1 * 1.000 ST
Product Details & Documents
Caps for 20 ml glass headspace crimp top round bottom vials which have a 23 mm OD and 75,5 mm height.
Spécifications
  • Désignation:
    PTFE coated silicone cap, pre-assembled
  • Seq:
    06