Closures for headspace vials
Aluminium coated silicone cap/septa/spring, unassembled
N° de catalogue PERCB0104243
Fournisseur: PerkinElmer
Aller à :
Product Details & Documents
Caps for 20 ml glass headspace crimp top round bottom vials which have a 23 mm OD and 75,5 mm height.
Spécifications
- Désignation:Aluminium coated silicone cap/septa/spring, unassembled
- Seq:04