Affinity protein expression and purification systems
Calmodulin affinity resin, Calmodulin binding peptide (CBP) affinity resin, 10 mL
N° de catalogue STRA214303
Affinity protein expression and purification systems
N° de catalogue STRA214303
Numéro CAS:
Aller à :
Arrêté
Product Details & Documents
The affinity protein expression and purification system provides a simple, gentle and effective method for high-level protein expression and purification.
- pCAL vectors derived from T7 RNA polymerase-based pET vectors
- Novel purification tag combines small size, gentle elution and high-affinity binding
- Easy tag removal resulting in native protein
Livraison: The complete system includes pCAL expression vectors containing a calmodulin-binding peptide tag and calmodulin affinity resin (available separately).
Attention: For research use only. Not for use in diagnostic procedures.
Spécifications
Spécifications
- Cdt:10 mL
- Désignation:Calmodulin affinity resin
- Details:Calmodulin binding peptide (CBP) affinity resin