Microplaques, à 1536 puits, Nunc™
Microplaque à 1536 puits, transparente, incolore, 1536, -
N° de catalogue 732-2711
Fournisseur: Thermo Fisher Scientific
Microplaques, à 1536 puits, Nunc™
N° de catalogue 732-2711
Fournisseur: Thermo Fisher Scientific
Aller à :
Product Details & Documents
PS, non treated, without lid. Designed for ultra-high throughput screening applications.
- Standard microplate footprint
- Available non treated in black, white or clear
- Low profile lid available separately
Working range: 1 to 10 µl.
Spécifications
- Nb de puits:1536
- Stérile:-
- Couleur:incolore
- Désignation:Microplaque à 1536 puits, transparente
- Cat. No.:732-2711
- Certification environnementale:NAR Certificate of Retirement
- Environmentally Preferable:Low Manufacturing Impact