pSTBlue-1 AccepTor™ vector kit
pSTBlue-1 AccepTor™ vector kit
pSTBlue-1 AccepTor™ Vector Kit, 20 Tests
N° de catalogue 70595-3
pSTBlue-1 AccepTor™ vector kit
N° de catalogue 70595-3
Numéro CAS:  
pSTBlue-1 AccepTor™ vector kit
pSTBlue-1 AccepTor™ Vector Kit, 20 Tests
Numéro CAS:  
Prix
504,00 €
Connexion Pour la disponibilité
1 * 20 Tests
Product Details & Documents
The pSTBlue-1 AccepTor™ vector kits are designed to simplify the PCR cloning process by using non-proofreading thermostable DNA polymerases (eg. KOD XL polymerase and native and recombinant Taq polymerases), which leave single 3'-dA overhangs on the reaction products. The AccepTor™ vectors enable direct ligation by virtue of single 3'-dU overhangs that anneal efficiently with 3'-dA extensions on PCR products. The dU residues are converted to dT residues in vivo following transformation.
Delivery information: AccepTor™ vectors are supplied in a ready-to-ligate format.
Spécifications
Spécifications
  • Cdt:
    20 Tests
  • Désignation:
    pSTBlue-1 AccepTor™ Vector Kit