Extraction thimbles
Extraction thimbles
33×90 mm, 1,5 mm
N° de catalogue 515-8929
Extraction thimbles
N° de catalogue 515-8929
Extraction thimbles
Extraction thimbles
33×90 mm, 1,5 mm
Prix
137,00 €
Connexion Pour la disponibilité
1 * 25 ST
Product Details & Documents
Grade 603 standard cellulose extraction thimbles, made of high quality cellulose, are available in a range of sizes suitable for use in thimble holders and automated extraction apparatus to ensure optimal fit.
Spécifications
  • Ø int.×h:
    33×90 mm
  • Seq:
    16
  • Épaisseur:
    1,5 mm