Extraction thimbles
28×100 mm, 1,5 mm
N° de catalogue 515-8918
Fournisseur: Whatman products (Cytiva)
Aller à :
Product Details & Documents
Grade 603 standard cellulose extraction thimbles, made of high quality cellulose, are available in a range of sizes suitable for use in thimble holders and automated extraction apparatus to ensure optimal fit.
Documents
WHAT10350211_TDS_EN
Fiche technique
Spécifications
- Ø int.×h:28×100 mm
- Seq:11
- Épaisseur:1,5 mm