Extraction thimbles
Extraction thimbles
25×60 mm, 1,5 mm
Catalog # 515-0080
Extraction thimbles
Catalog # 515-0080
Extraction thimbles
Extraction thimbles
25×60 mm, 1,5 mm
Price
111,00 €
Log in for availability
1 * 25 ST
Product Details & Documents
Grade 603 standard cellulose extraction thimbles, made of high quality cellulose, are available in a range of sizes suitable for use in thimble holders and automated extraction apparatus to ensure optimal fit.
Specifications
  • Ø int.×h:
    25×60 mm
  • Seq:
    04
  • Épaisseur:
    1,5 mm