Micro control valves
Brass micro control valve (BMCV-1), "L" single outlet, connection 1,6 mm, temperature 100 °C, 700 psi pressure, flow rate 0 - 2000 ml/min
N° de catalogue SGEA1236282
Fournisseur: Trajan Scientific and Medical
Aller à :
Product Details & Documents
Micro control valves provide precision metering and on-off control of gas flows. They are designed specifically for chromatography applications and used at elevated temperatures from high vacuum to medium pressure.
Actuated remotely with a toggle switch or solenoid valve, and a 50 to 55 psi air supply.
Spécifications
- Désignation:Brass micro control valve (BMCV-1), "L" single outlet, connection 1,6 mm, temperature 100 °C, 700 psi pressure, flow rate 0 - 2000 ml/min
Accessories
Informations sur le produit
- N° de catalogueDésignationDisponibilitéPrix
Informations sur le produit
- N° de catalogueDésignationDisponibilitéPrix
Informations sur le produit
- N° de catalogueDésignationDisponibilitéPrix