Micro control valves
Manual on/off valve, pneumatic (MOVPTL), "T" dual outlet, stem length 100 mm, connection 1,6 mm, temperature 300 °C, vacuum rated, 700 psi pressure
N° de catalogue SGEA1236104
Fournisseur: Trajan Scientific and Medical
Aller à :
Product Details & Documents
Micro control valves provide precision metering and on-off control of gas flows. They are designed specifically for chromatography applications and used at elevated temperatures from high vacuum to medium pressure.
Actuated remotely with a toggle switch or solenoid valve, and a 50 to 55 psi air supply.
Spécifications
- Désignation:Manual on/off valve, pneumatic (MOVPTL), "T" dual outlet, stem length 100 mm, connection 1,6 mm, temperature 300 °C, vacuum rated, 700 psi pressure
Accessories
Informations sur le produit
- N° de catalogueDésignationDisponibilitéPrix
Informations sur le produit
- N° de catalogueDésignationDisponibilitéPrix
Informations sur le produit
- N° de catalogueDésignationDisponibilitéPrix