Base materials
Peptone from meat, 250 g
N° de catalogue SERA48620.04
Aller à :
Arrêté
Product Details & Documents
High quality source of peptides and amino acids produced by enzymatic digestion.
- Suitable as nutrient for laboratory media and industrial fermentation
Spécifications
Spécifications
- Cdt:250 g
- Désignation:Peptone from meat