Flat flange lids, centre neck, DURAN®
bride plate rodée DN 100, 138 mm, 1 col, NS 29/32 (centre)
N° de catalogue SCOT243984605
Fournisseur: DWK Life Sciences
Flat flange lids, centre neck, DURAN®
N° de catalogue SCOT243984605
Fournisseur: DWK Life Sciences
Aller à :
Product Details & Documents
DURAN®, borosilicate glass 3.3, clear.
- Very good chemical resistance
- High temperature resistance
- Minimal thermal expansion giving relatively high resistance to temperature changes
Spécifications
- Ø ext.:138 mm
- Type:1 col
- Avec:bride plate rodée DN 100
- Col:NS 29/32 (centre)