Sample vials, clear, with screw thread, KIMBLE®
Sample vials, clear, with screw thread, KIMBLE®
N° de catalogue 391-6132
Fournisseur:   DWK Life Sciences
Sample vials, clear, with screw thread, KIMBLE®
N° de catalogue 391-6132
Fournisseur:  DWK Life Sciences
Sample vials, clear, with screw thread, KIMBLE®
Sample vials, clear, with screw thread, KIMBLE®
Prix
91,20 €
Connexion Pour la disponibilité
1 * 100 ST
Product Details & Documents
Screw threaded sample vials made from soda-lime glass, with white PE caps unattached.