Hexane (mélange d'isomères), technical
Flacon Verre, 2,5 L
: Hexanes, Isohexane
- :92112-69-1
- :H₃C(CH₂)₄CH₃
- :MFCD00009520
- :86.18 g/mol
- :69 °C
- :–95 °C
- :0,659 g/cm³
- :–22 °C
- :Température ambiante
- :295-570-2
- :1208
- :3,II
- :01-2119484651-34
- Appearance:Clear liquid
- Color scale:≤10 APHA
- Residue after evaporation:≤0.002 %
- Type de conditionnement:Flacon Verre
- Cdt:2,5 L
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



