Test sets
Test set II for inorganic gases
N° de catalogue 151-0177
Fournisseur: Dräger
Aller à :
Product Details & Documents
These test sets allow for the semi quantitative measurements of five different gases at the same time.
Spécifications
- Désignation:Test set II for inorganic gases
- Seq:02
- Cat. No.:151-0177
- Durée de mesure:40 s
- Modèle:Test set II for inorganic gases