Fluorescéine sodium, TECHNICAL
Seau (plastique), 5 kg
: 3',6'-Dihydroxy-3H-spiro[isobenzofuran-1,9'-xanthen]-3-one, disodium salt, Acid Yellow 73, Yellow No. 8, Fluorescein sodium salt, 2-(6-hydroxy-3-oxo-(3H)-xanthen-9-yl)benzoic acid disodium salt, Disodium 2-(3-oxo-6-oxidoxanthen-9-yl)benzoate
- :518-47-8
- :C₂₀H₁₀Na₂O₅
- :MFCD00167039
- :376.28 g/mol
- :13,04185
- :> 300 °C
- :1,601 g/cm³ (20 °C)
- :Température ambiante
- :208-253-0
- Appareance:Passes test
- IR Spectrum:Passes test
- UV Spectrum:Passes test
- Absorption max. (buffer pH 8,0):490 - 492 nm
- Spec.absorptivity(Lm;0,005g/l;pH8,0;dry):Min. 1200
- Assay:Min. 80%
- Loss on drying (110°C) at manufacturing:Max. 10 %
- Type de conditionnement:Seau (plastique)
- Cdt:5 kg
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



