Tert-Butyle methyle oxyde 99%, pur
Flacon Verre, 2,5 L
: MTBE, 2-Methoxy-2-methylpropane, Methyl-t-Butyl Ether
- :1634-04-4
- :(CH₃)₃COCH₃
- :MFCD00008812
- :88.15 g/mol
- :13,06059
- :54…56 °C (760 mmHg)
- :–110 °C
- :0,7404 g/cm³
- :–28 °C
- :Température ambiante
- :216-653-1
- :2398
- :3,II
- Appearance:Clear colorless Liquid
- Infrared spectrum:Conforms
- Refractive index:1.3675 to 1.3695 (20°C, 589 nm)
- Water:≤0.01 % (Coulometric)
- GC:≥98.5 %
- Type de conditionnement:Flacon Verre
- Cdt:2,5 L



