Thermometer test caps
TX26L, test cap, Lumberg type, +70 °C
N° de catalogue 620-0380
Fournisseur: COMARK
Aller à :
Product Details & Documents
Certified test capsules can be used for quick checking of the thermometer's function against specifications. They are connected to the thermometer instead of a sensor and consist of a resistor that corresponds exactly to a given temperature.
Spécifications
- Plage de mesure:+70 °C
- Désignation:TX26L, test cap, Lumberg type