1-Méthylimidazole 99%
Flacon verre pour solides, 100 g
Supplier: Thermo Fisher Scientific
1-Méthylimidazole 99%
Catalog # ACRO126991000
Supplier: Thermo Fisher Scientific
CAS Number: 616-47-7
Jump to:
Product Details & Documents
Synonyms: N-Methylimidazole, 1-Methyl-1H-imidazole, N-Methyl-1H-imidazole, NMI
- CAS Number:616-47-7
- Formula:C₄H₆N₂
- MDL Number:MFCD00005292
- Molecular Weight:82.11 g/mol
- Boiling Pt:198 °C (1013 hPa)
- Melting Pt:–2 °C
- Density:1,03 g/cm³
- Flash Pt:92 °C
- Storage Temperature:Température ambiante
- EINECS:210-484-7
- UN:2922
- ADR:8,II
Documents
Specifications
Specification Test Results
- Appearance:Clear colorless to yellow Liquid (May discolor to yellow on storage)
- Infrared spectrum:Conforms
- Refractive index:1.4955 to 1.4975 (20°C, 589 nm)
- Water:≤0.5 % (K.F.)
- GC:≥98.5 %
- Color scale:≤15 APHA
Specifications
- Type de conditionnement:Flacon verre pour solides
- Cdt:100 g
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



