Saisie de commande
Belgium
Lien pour commandesContactUsLinkComponent
Affinity chromatography columns, HiTrap™ Benzamidine FF

Affinity chromatography columns, HiTrap™ Benzamidine FF

HiTrap™ Benzamidine FF (High Sub), PP, 90 µm, 7×25 mm, 1 ml, <4 ml/min
N° de catalogue 17-5143-02
Fournisseur:   Cytiva
Affinity chromatography columns, HiTrap™ Benzamidine FF
N° de catalogue 17-5143-02
Fournisseur:  Cytiva
Affinity chromatography columns, HiTrap™ Benzamidine FF
Affinity chromatography columns, HiTrap™ Benzamidine FF

HiTrap™ Benzamidine FF (High Sub), PP, 90 µm, 7×25 mm, 1 ml, <4 ml/min

Prix
226,00 €
Connexion Pour la disponibilité
1 * 2 mL
Product Details & Documents

HiTrap Benzamidine FF (high sub) are prepacked with Benzamidine Sepharose 4 Fast Flow (high sub) and designed specifically for the removal and/or purification of serine proteases such as thrombin.

  • Effective removal of thrombin and factor Xa after tag cleavage of recombinant proteins
  • Removal of serine proteases is easily done directly from serum, monoclonal cell supernatants and bacterial lysates
  • High binding capacity
  • It can be operated using syringe, peristaltic pump or liquid chromatography system such as ÄKTA design or FPLC system

HiTrap Benzamidine FF (high sub) are prepacked 1 ml and 5 ml columns for convenient, one-step removal and/or purification of trypsin, trypsin-like serine proteases, and zymogens including urokinase and prekallikrein.

The medium, Benzamidine Sepharose 4 Fast Flow (high sub), is also available as lab packages and is an excellent choice for scale-up.

Spécifications
  • Débit:
    <4 ml/min
  • Granulométrie:
    90 µm
  • Capacité:
    1 ml
  • Désignation:
    HiTrap™ Benzamidine FF (High Sub)
  • Seq:
    02
  • Matériau de colonne:
    PP
  • Dimensions du lit:
    7×25 mm
  • Plage de pH:
    2 - 8
  • Cat. No.:
    17-5143-02
  • Stockage:
    2 to 8 °C, 20% Ethanol, 0,05 M Sodium Acetate, pH 4,0
  • Pression max.:
    5 bar
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.